Learn More
STEMCELL Technologies EasySep™ EasyStand™ with Magnet
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
- The EasySep™ EasyStand™ facilitates the separation of one or more samples at one time
- Each EasySep™ EasyStand™ can hold a single EasySep™ Magnet (Catalog #18000), and a simple system links up to 6 individual EasySep™ EasyStands™ together
- After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes
Specifications
Specifications
| For Use With (Application) | Cell Isolation |
| Quantity | 1 Ea. |