Learn More
STEMCELL Technologies EasySep™ EasyStand™ with Magnet
Product Code. 15775512
Encompass_Preferred
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. 15775512
Brand: STEMCELL Technologies 18131
One linkable stand and one EasySep™ Magnet
  • The EasySep™ EasyStand™ facilitates the separation of one or more samples at one time
  • Each EasySep™ EasyStand™ can hold a single EasySep™ Magnet (Catalog #18000), and a simple system links up to 6 individual EasySep™ EasyStands™ together
  • After each round of magnetic separation, the supernatants from all samples are poured off simultaneously into the receiving tubes
Specifications
For Use With (Application) Cell Isolation
Quantity 1 Ea.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.