Learn More
STEMCELL Technologies EasySep™ Direct Human CD4+ T Cell Isolation Kit
Click to view available options
Quantity:
100 mL whole blood
Unit Size:
Each
Description
- Isolate CD4+ T cells directly from human whole blood by immunomagnetic negative selection
- Desired cells undergo minimal manipulation and are untouched and highly purified
- This product labels RBCs, platelets and unwanted cells with antibody complexes and magnetic particles
- The magnetically labeled RBCs, platelets and non-target cells are separated from the untouched CD4+ T cells in an EasySep™ magnet and poured or pipetted into a new tube
- Isolated cells are available for use in downstream uses such as flow cytometry, functional assays or culture
Specifications
Specifications
| Cell Type | T Cells, T Cells, CD4+ |
| For Use With (Application) | Cell Isolation |
| Quantity | 100 mL whole blood |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.