missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Direct Human CD4+ T Cell Isolation Kit for RoboSep™

Product Code. 17197941
Encompass
Click to view available options
Quantity:
100 mL whole blood
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17197941

Brand: STEMCELL Technologies 19662RF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips

  • Isolate CD4+ T cells directly from human whole blood by immunomagnetic negative selection
  • Desired cells undergo minimal manipulation and are untouched and highly purified
  • This product labels RBCs, platelets and unwanted cells with antibody complexes and magnetic particles
  • The magnetically labeled RBCs, platelets and non-target cells are separated from the untouched CD4+ T cells in an EasySep™ magnet and poured or pipetted into a new tube
  • ​Isolated cells are available for use in downstream uses such as flow cytometry, functional assays or culture

Specifications

Cell Type T Cells, T Cells, CD4+
For Use With (Application) Cell Isolation
Quantity 100 mL whole blood
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.