Learn More
BRAND™ EASYCAL™ 5.0 Server License
Product Code. 17841252 Shop All Brand Products
Change view
Click to view available options
For Use With (Application):
Calibration for Large Laboratories
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
17841252 Calibration for Large Laboratories Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17841252 Supplier BRAND™ Supplier No. 708414
Estimated Shipment: 20-11-2026
Server license of the EASYCAL 5.0 calibration software with free updates.
The EASYCAL™ 5 calibration software facilitates the monitoring of Liquid Handling instruments (like piston-stroke devices such as pipettes, dispensers, burettes, and manual dispensers), as well as volumetric instruments made of glass or plastic, based on GLP/GMP and DIN EN ISO 9001. EASYCAL™ 5 software can be used for BRAND instruments as well as instruments from other manufacturers.
  • Automatically calculates and compares all values against the tolerances from current standards or individually-defined limits (tolerances of numerous devices and the interface settings of over 100 test devices (e.g. balances) are already stored for you)
  • Choose between a stand-alone license for working on one workstation or a client/server version for parallel, distributed work on multiple workstations
    • Stand-alone license is recommended for small labs where calibration is completed by one person
    • Client/server version and its floating licenses are ideal for larger labs where calibration will be carried out at multiple workstations
  • For testing of liquid handling devices and volumetric instruments made of glass and plastic according to standards such as ISO 8655, ISO 4787, and others
  • Open software, suitable for all volumetric instruments - from any manufacturer
  • Extensive library with device specifications from many well-known manufacturers can be expanded and modified by the user to help you get started and minimize data entry time
  • Quickly and easily search and find the owner, test history, and next test date
  • Continuous control of current actual state during testing by graphical representations and real time calculation of statistical values
  • Reminder function for upcoming tests with automatic email notifications sent to device owner
  • Integration of address data of your customers and suppliers in a business partner database
  • User management with user roles (e.g., inspector, supervisor, and system administrator) and access restriction to EASYCAL functions
  • Dual-control principle for the release of critical data such as test plans, calibration orders before certificate printing, and device specification
  • RS-232 interface connection for measuring devices such as scales, thermometers, barometers, and hygrometers with automatic data transmission of the measured values
  • Customize certificates and audit trails to your needs in the integrated certificate editor
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Software
For Use With (Application) Calibration for Large Laboratories