missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ E2F6 (Human) Recombinant Protein
Human E2F6 full-length ORF ( AAH08348, 1 a.a. - 281 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001876-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSQQRPARKLPSLLLDPTEETVRRRCRDPINVEGLLPSKIRINLEDNVQYVSMRKALKVKRPRFDVSLVYLTRKFMDLVRSAPGGILDLNKVATKLGVRKRRVYDITNVLDGIDLVEKKSKNHIRWIGSDLSNFGAVPQQKKLQEELSDLSAMEDALDELIKDCAQQLFELTDDKENERLAYVTYQDIHSIQAFHEQIVIAVKAPAETRLDVPAPREDSITVHIRSTNGPIDVYLCEVEQGQTSNKRSEGVGTSSSESTHPEGPEEEENPQQSEELLEVSN
Specifications
AAH08348 | |
E2F transcription factor 6 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
E2F6 | |
GST |
1876 | |
Wheat germ expression system | |
10 μg | |
E2F-6, MGC111545 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |