Learn More
Bochem™ E-Poli Test Tube Stands
Product Code. 10410163
Click to view available options
Array:
10 x 10
2 x 10
2 x 12
3 x 12
4 x 12
4 x 6
Unit Size:
Each
Product Code. 10410163
Brand: Bochem™ 10175
Specifications
Product Type Test Tube Rack
Array 4 x 12
Holds Tubes (16 mm)
Material 18/10 E-Poli
Positions 48
Diameter (Metric) Opening 16 mm