Learn More
(E,E)-4,8,12-Trimethyl-1,3,7,11-tridecatetraene, TRC
Specifications
Specifications
| Chemical Name or Material | (E,E)-4,8,12-Trimethyl-1,3,7,11-tridecatetraene |
| CAS | 62235-06-7 |
| Purity Grade Notes | GC |
| Molecular Formula | C16H26 |
| Quantity | 100 mg |
| Synonym | (3E,7E)-4,8,12-Trimethyl-1,3,7,11-tridecatetraene, 4,8,12-Trimethyl-1,3E,7E,11-tridecatetraene |
| InChI Formula | InChI=1S/C16H26/c1-6-9-15(4)12-8-13-16(5)11-7-10-14(2)3/h6,9-10,13H,1,7-8,11-12H2,2-5H3/b15-9+,16-13+ |
| SMILES | O[C@H]1C[C@@H](CCC1)NC(=O)c2ccccc2 |
| Molecular Weight (g/mol) | 218.38 |
| Percent Purity | >95 |