Learn More
(E)-Acetylacrylic acid, TRC
Product Code. 30308895
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30308895 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30308895 Supplier Toronto Research Chemicals Supplier No. TRCA169435500MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (E)-Acetylacrylic acid
CAS 2833-28-5
Molecular Formula C5H6O3
Quantity 500 mg
Synonym (2E)-4-Oxo-2-pentenoic Acid, (2E)-4-Oxopent-2-enoic Acid, (E)-3-Acetylpropenoic Acid, (E)-4-Oxopent-2-enoic Acid, (E)-4-Oxopent-2-enoic Acid, trans-3-Acetylacrylic Acid, trans-4-Oxo-2-pentenoic Acid, trans-beta-Acetylacrylic Acid
InChI Formula InChI=1S/C5H6O3/c1-4(6)2-3-5(7)8/h2-3H,1H3,(H,7,8)/b3-2+
SMILES CC(=O)\C=C\C(=O)O
IUPAC Name (E)-4-oxopent-2-enoic acid
Molecular Weight (g/mol) 114.1
Formula Weight 114.0317