Learn More
3M™ E-A-R™ Ultrafit™ Ear Plugs, Corded
Click to view available options
Color:
Green
Yellow
Yellow, Blue
Noise Reduction Rating (NRR):
14 db
20 db
32 db
Material:
Elastomeric Polymer
Silicone Rubber
TPE
Thermoplastic Elastomer
Quantity:
200/Pk.
50/Pk.
Unit Size:
50 pairs
Pack of 200
Descrizione
- Triple-flange pliable plug effectively fits most ear canal sizes
- Reduces noise levels by 14-32 dB
- Ideal for dirty environments as no roll down required, simply push them into your ears for hygienic protection
- Clean easily with soapy water and reuse again and again, helping decrease waste
- Removable, reusable cord attachment – convenient if you move in and out of noisy environments
- Available with a storage case to keep plugs clean and together
Specifica
Specifica
| Color | Green |
| Certifications/Compliance | CE Approved |
| Quantity | 200/Pk. |
| Noise Reduction Rating (NRR) | 14 db |
| Product Type | Ear Plugs |
| Material | Silicone Rubber |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto