missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ E 7820

Product Code. 18739236
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18739236 50 mg 50mg
18717226 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18739236 Supplier R&D Systems™ Supplier No. 6785/50
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. α2 integrin inhibitor; anti-angiogenic

E 7820 is a α2 integrin inhibitor. Inhibits angiogenesis in vitro (IC50 = 0.11 - 0.25 μM). Also inhibits growth of human colorectal cancer cell lines. Enhances efficacy of erlotinib in a non-small-cell lung cancer xenograft model. Also acts as a molecular glue to induce proteosomal degradation of mRNA splicing factor RBM39.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Cyano-N-(3-cyano-4-methyl-1 H-indol-7-yl)benzenesulfonamide
CAS 289483-69-8
Quantity 50 mg
Product Line Tocris
Target Integrin Receptor Inhibitors
Molecular Formula C17H12N4O2S
Formula Weight 336.37
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.