missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ SARS-CoV-2 Spike IgM ELISA Kit

Product Code. p-9582002
Click to view available options
Quantity:
96 reactions
Unit Size:
Pack of 96
This item is not returnable. View return policy

Product Code. 17561825

Brand: Invitrogen™ 18010D

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

The Dynabeads™ SARS-CoV-2 Spike IgM ELISA Kit is a solid‐phase sandwich enzyme‐linked immunosorbent assay (ELISA) designed to detect and quantify the level of human SARS-CoV-2 IgM from serum and plasma samples. The product can be used both for qualitative and quantitative assays.

  • A solid‐phase sandwich enzyme‐linked immunosorbent assay (ELISA) designed to detect and quantify the level of human SARS-CoV-2 IgM from serum and plasma samples
  • The steps required for use are simply to pre-dilute the reagents, fill the wells of the KingFisher plastic plates, and push start on the instrument
  • After the run, the sample is transferred to a plate compatible with the absorbance reader and the OD is read at 450 nm
  • A DynaMag-96 well magnet can be used while still taking advantage of the faster binding kinetics of magnetic beads compared to the more traditional plate-bound ELISAs
  • Kit contains all buffers required, as well as calibrator antibodies, HRP-conjugated detection antibodies, substrate solution, beads, and stop solution sufficient for one 96-well plate
  • The intra-assay CV value is typically < 5% (range 2.4-3.9%), while the inter-assay CV is < 15% (range 3.1-14.4%)
  • Assay can be used both for qualitative and quantitative assays
  • One of the fastest ELISA assays for SARS-CoV-2 serology
  • Dynabeads facilitated automation limits hands-on time and improves reproducibility by reducing most operating errors
  • For research use only, not for use in diagnostic procedures
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 9001
Content And Storage • Dynabeads; SARS-CoV-2 Spike, 3 mL (1 mg/mL)
• Human SARS-CoV-2 IgM High Control antibody, 1 vial (lyophilized)
• Human SARS-CoV-2 IgM Low Control antibody, 1 vial (lyophilized)
• Human SARS-CoV-2 IgM Calibrator (medium control) antibody, 1 vial (lyophilized)
• Wash buffer (20X concentrated), 50 mL
• Assay buffer (20X concentrated), 50 mL
• Substrate solution (tetramethylbenzidine), 15 mL
• Stop solution, 15 mL
• Human SARS-CoV-2 IgM HRP Conjugate, 0.12 mL

Store at 2–8°C.

Detection Method Absorbance at 450 nm
Product Type SARS-CoV-2 ELISA IgM Kit
For Use With (Application) Binding of human IgM towards SARS-CoV-2 (Spike trimer)
For Use With (Equipment) KingFisher™ Flex or Apex Instruments, DynaMag™-96 Side Skirted, Magnetic 96-Well Separator, ELISA Plate Reader (450 nm)
Quantity 96 reactions
Shipping Condition Wet ice (2° to 8°C)
Diameter (Metric) Bead 2.7 μm
Material Monosized, superparamagnetic polystyrene magnetic beads
High-throughput Compatibility Yes, on KingFisher™ Flex or Apex instruments
Show More Show Less

For research use only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.