missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Dynabeads™ SARS-CoV-2 Spike IgM ELISA Kit
Click to view available options
Quantity:
96 reactions
Unit Size:
Pack of 96
Description
- A solid‐phase sandwich enzyme‐linked immunosorbent assay (ELISA) designed to detect and quantify the level of human SARS-CoV-2 IgM from serum and plasma samples
- The steps required for use are simply to pre-dilute the reagents, fill the wells of the KingFisher plastic plates, and push start on the instrument
- After the run, the sample is transferred to a plate compatible with the absorbance reader and the OD is read at 450 nm
- A DynaMag-96 well magnet can be used while still taking advantage of the faster binding kinetics of magnetic beads compared to the more traditional plate-bound ELISAs
- Kit contains all buffers required, as well as calibrator antibodies, HRP-conjugated detection antibodies, substrate solution, beads, and stop solution sufficient for one 96-well plate
- The intra-assay CV value is typically < 5% (range 2.4-3.9%), while the inter-assay CV is < 15% (range 3.1-14.4%)
- Assay can be used both for qualitative and quantitative assays
- One of the fastest ELISA assays for SARS-CoV-2 serology
- Dynabeads facilitated automation limits hands-on time and improves reproducibility by reducing most operating errors
- For research use only, not for use in diagnostic procedures
Specifications
Specifications
| Certifications/Compliance | ISO 9001 |
| Content And Storage | • Dynabeads; SARS-CoV-2 Spike, 3 mL (1 mg/mL) • Human SARS-CoV-2 IgM High Control antibody, 1 vial (lyophilized) • Human SARS-CoV-2 IgM Low Control antibody, 1 vial (lyophilized) • Human SARS-CoV-2 IgM Calibrator (medium control) antibody, 1 vial (lyophilized) • Wash buffer (20X concentrated), 50 mL • Assay buffer (20X concentrated), 50 mL • Substrate solution (tetramethylbenzidine), 15 mL • Stop solution, 15 mL • Human SARS-CoV-2 IgM HRP Conjugate, 0.12 mL Store at 2–8°C. |
| Detection Method | Absorbance at 450 nm |
| Product Type | SARS-CoV-2 ELISA IgM Kit |
| For Use With (Application) | Binding of human IgM towards SARS-CoV-2 (Spike trimer) |
| For Use With (Equipment) | KingFisher™ Flex or Apex Instruments, DynaMag™-96 Side Skirted, Magnetic 96-Well Separator, ELISA Plate Reader (450 nm) |
| Quantity | 96 reactions |
| Shipping Condition | Wet ice (2° to 8°C) |
| Diameter (Metric) Bead | 2.7 μm |
| Material | Monosized, superparamagnetic polystyrene magnetic beads |
| Show More |
For research use only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction