missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ DyLight™ 800 Maleimide
Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Thermo Scientific™ Sulfhydryl-Reactive DyLight Dyes are maleimide-activated DyLight Fluors for fluorescent labeling of antibodies and other proteins to be used as probes for cellular imaging and other fluorescence detection methods.
The Thermo Scientific™ DyLight™ Dyes have absorption spectra ranging from 350 to 770nm. These reagents fluoresce over a broad pH range, are more intense than Alexa Fluor™ or Cy™ Dyes in many applications and match the output wavelengths of common fluorescence instrumentation. Additionally, the water solubility of the DyLight Reagents allows a high fluor-to-protein ratio without precipitation during conjugation.
The sulfhydryl-reactive dyes contain maleimide groups that react predominantly with free –SH groups at pH 6.5-7.5, forming a stable thioether bond. At pH 7, the maleimide group is ∼1000 times more reactive toward a free sulfhydryl than to an amine.1At pH values > 7.5, reactivity toward primary amines increases and hydrolysis of the maleimide group can occur.
Highlights:
Applications:
The sulfhydryl-reactive dyes contain maleimide groups that react predominantly with free –SH groups at pH 6.5-7.5, forming a stable thioether bond. At pH 7, the maleimide group is ∼1000 times more reactive toward a free sulfhydryl than to an amine.1At pH values > 7.5, reactivity toward primary amines increases and hydrolysis of the maleimide group can occur.
Highlights:
- Specific — maleimide-activated dye labels proteins and other molecules at reduced sulfhydryls (-SH)
- Efficient labeling methods — well-characterized chemistry and optimized protocols provide for reliable, high-quality labeling
- Optimized antibody labeling procedure —complete protocol for IgG reduction and labeling and calculating the labeling efficiency
Applications:
- Antibody labeling for immunofluorescence applications, including immunocytochemistry (ICC), immunohistochemistry (IHC), Western blotting and ELISA assay
- Target macromolecule labeling for in vitro and in vivo fluorescent detection strategies
Specifications
Specifications
| Description | DyLight 800 Maleimide |
| Quantity | 1 mg |
| Product Type | Dye |
| Solubility | DMF (Dimethylformamide) |
| Content And Storage | Store desiccated at -20°C. |
| Chemical Reactivity | Thiol |
| Reactive Moiety | Maleimide |
| Label or Dye | DyLight™ 800 |
| Label Type | DyLight Fluors |
| Product Line | DyLight™ |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction