Learn More
DWK Life Sciences DURAN™ Volumetric Flask, Class A, individual certificate
Product Code. p-7189233 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
1 mL
10 mL (Wide neck)
100 mL (Stopper Size 12/21)
100 mL (Stopper Size 14/23)
1000 mL
1000 mL (Wide neck)
2 mL
20 mL
200 mL
2000 mL
25 mL
250 mL
5 mL (Wide neck)
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 2
Product Code. 15270876
Brand: DWK Life Sciences 246790151
DURAN™ volumetric flasks are used for the accurate measurement of specific quantities of liquids. They are volumetric analysis aids in quantitative laboratory work. These volumetric flasks are certified item by item and are marked with an individual number.
Volumetric flasks are available in two different accuracy Classes: Class A and Class B. The two Classes differ in the accuracy of measurement with Class A being the highest accuracy, and Class B is approximately half that of Class A. The volumetric flasks with accuracy Class A have a blue print for clear glass products and a white print for amber versions. Volumetric flasks which are numbered individually, are supplied with an individual certificate. The individual number is permanently laser-etched onto the base of the volumetric flask and is entered on the corresponding certificate. The volume measured for the corresponding volumetric flask, the measurement uncertainty and the day of issue are documented on this certificate. The individual number is a consecutive number and comprises three letters and a four digit number, for example: AAA-0001.
  • Calibration based on the poured in volume (In) at a + 20°C reference temperature
  • Outstanding precision during volumetric measurement and high-quality process stability
  • Thanks to the exact processing and precisely calibrated scales, they permit the highly accurate determination and measurements of volume
  • Supplied with ergonomic polyethylene stopper for easy and safe tightening or removal
  • The volume content tolerances conform to accuracy Class A
  • Fired-on, highly durable blue print
  • With printed ring mark for better visibility
  • Influence of undesirable factors is kept to an absolute minimum
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Base 13 mm
Capacity (Metric) 1 mL
Material Borosilicate Glass (3.3), Polyethylene
For Use With (Application) Preparation and storage of standard solutions; Measuring liquid amounts; Diluting solutions; Mixing several components; Measurement analysis; Volumetric measurement; To ensure a long service life for your volumetric glassware and to exclude possible volume changes, these glass products should not be heated above + 250°C in drying cabinets or sterilisers
No. per Pack 2/Pk.
Tolerance ± 0.025 mL