Learn More
DWK Life Sciences DURAN™ Triple-Neck Round Bottom Flask, with standard ground joint, parallel side necks
Product Code. p-7189494 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Standard Taper Joint:
Parallel side necks NS 14/23
Parallel side necks NS 19/26
Parallel side necks NS 29/32
Unit Size:
Each
Product Code. 10171781
Brand: DWK Life Sciences 241854401
Only null left
Add to basket
Add to basket
Multi-neck pellets are used for distillation, fraction and reactions of all kinds.
For distillation, fraction and reaction, mainly multi-neck flasks (two or three neck flasks) are used to be able to install attachments, columns, thermometers, dropping funnels, boiling capillaries, etc. as required.
  • Tight closure due to standard ground joint
  • Parallel side necks
  • Depending upon the application, accessories, columns, thermometers, dropping funnels, boiling capillaries, etc. can be fitted
  • The geometry permits very uniform heating
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Base 105 mm
Capacity (Metric) 500 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Distillation; Extraction; Connection to other glass vessels; Heating; Suitable for heating baths and mantles; Volume sizes 250 mL (NS 24/29 and NS 29/32), 500 mL (NS 24/29), 500 mL (NS 29/32), 1000 mL (NS 29/32) and 2000 mL (NS 29/32) are not according to DIN ISO
No. per Pack 1/Pk.
Product Type Non-DIN/ISO size
Standard Taper Joint Parallel side necks NS 19/26