Learn More
DWK Life Sciences DURAN™ Triple-Neck Round Bottom Flask, with standard ground joint, angled side necks
Product Code. 10749211 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Standard Taper Joint:
Center neck NS 24/29
Center neck NS 29/32
Unit Size:
Each
Product Code. 10749211
Brand: DWK Life Sciences 241885306
Multi-neck pellets are used for distillation, fraction and reactions of all kinds.
For distillation, fraction and reaction, mainly multi-neck flasks (two or three neck flasks) are used to be able to install attachments, columns, thermometers, dropping funnels, boiling capillaries, etc. as required.
  • Tight closure due to standard ground joint
  • angled side necks
  • Depending upon the application, accessories, columns, thermometers, dropping funnels, boiling capillaries, etc. can be fitted
  • The geometry permits very uniform heating
  • Fired-on, highly durable white print
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Base 131 mm
Capacity (Metric) 1000 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Distillation; Extraction; Connection to other glass vessels; Heating; Suitable for heating baths and mantles
No. per Pack 1/Pk.
Standard Taper Joint Center neck NS 24/29