Learn More
DWK Life Sciences DURAN™ Standard Socket, double, diamond-ground
Product Code. 10131411 Shop All DWK Life Sciences Products
Click to view available options
Joint Size:
18 mm
28 mm
34 mm
Unit Size:
Each
Pack of 50
Product Code. 10131411
Brand: DWK Life Sciences 286573309
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed for a rigid, self-supporting connection with lower requirements.
DURAN™ DG double sleeves are not ground according to DIN 12249 and represent the cost-effective alternative to DURAN™ cores.
  • Diamond cut (coarser cut quality, more roughness depth)
  • Cost-effective alternative to DURAN™ sleeves (not according to DIN)
  • Glass article with two sleeves enables material savings
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25/Pk.
Item Description Installation in glass apparatus and devices
Joint Size 34 mm
Thickness (Metric) Wall 2 mm