Description
DURAN™ slotted screen plates serve as support surfaces for the actual filter (e.g. paper or membrane filter), e.g. in the case of buechner funnels.
- As support for paper or membrane filters
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Suitable for coarse filtration without additional filter medium; Filtration |
| Quantity | 10 |
| Diameter (Metric) | 60 mm |