Learn More
DWK Life Sciences DURAN™ Single way Stopcock, complete with PTFE key
Shop All DWK Life Sciences Products
Click to view available options
Plug Bore:
2 mm
2.5 mm
4 mm
6 mm
8 mm
10 mm
Closure Size:
12.5 NS
14.5 NS
18.8 NS
21.5 NS
24 NS
29.2 NS
Unit Size:
Pack of 25
Pack of 5
Pack of 50
Description
DURAN™ single way stopcocks complete with PTFE valve plugs allow grease free operation. The 3-part tap safety catch secures the tap plug in the tap sleeve.
- Polished shut-off valve sleeve with PTFE valve plug for grease-free operation
- The 3-part tap safety catch secures the tap plug in the tap sleeve.
- Taper 1:10
- Glass articles for further processing
Specifications
Specifications
| Plug Bore | 2 mm |
| Diameter (Metric) | 8 mm |
| Material | Borosilicate Glass (3.3), PTFE |
| Autoclavable | Autoclavable |
| Closure Size | 12.5 NS |
| Width (Metric) | 1.5 mm |
| No. per Pack | 50/Pk. |
| For Use With (Application) | Installation in glass apparatus and devices |