Learn More
DWK Life Sciences DURAN™ Screw Thread Coupling, from PBT
Product Code. 10768351 Shop All DWK Life Sciences Products
Click to view available options
Thread Size:
GL 14
GL 18
GL 25
GL 32
GL 45
Unit Size:
Each
Product Code. 10768351
Brand: DWK Life Sciences 292260659
Only null left
Add to basket
Add to basket
The screw thread coupling is used to connect two threaded connections.
The screw thread coupling consists of two threaded caps, which are connected to each other via a flexible hose piece. This component enables a flexible connection of two glass components.
  • For flexible connection of two glass screw-thread connections with integral silicone seal (VQM)
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material PBT
For Use With (Application) Connection of glass threads
Packaging Quantity 1 Ea.
Thread Size GL 18