Learn More
Duran Duran Protect+ Media Bottle 10L GL45 Clear PU coated
Brand:  Duran 218058686
Duran Protect+ Media Bottle 10L GL45 Clear PU coated
Product Code. 30052745
  • ONLINE EXCLUSIVE PRICE
    607.00€
    586.15€ / Each
    Save 20.85€
    3% Off