Learn More
Duran Duran Protect+ Media Bottle 10L GL45 Amber PU coated
Brand:  Duran 218068687
Duran Protect+ Media Bottle 10L GL45 Amber PU coated
Product Code. 30052747
  • ONLINE EXCLUSIVE PRICE
    767.00€
    741.89€ / Each
    Save 25.11€
    3% Off
Only null left
Estimated Shipment: 17-02-2026