Learn More
DWK Life Sciences DURAN™ Premium Screw Cap, Colourless PFA, with PTFE-coated (Platinum cured) silicone cap liner
Product Code. p-7189205 Shop All DWK Life Sciences Products
Click to view available options
Thread Size (Metric):
GL 25
Unit Size:
Pack of 5
Product Code. 13177226
Brand: DWK Life Sciences 1129600
Edge
Screw bottle closure with cap liner sealing that offers higher temperature and chemical resistance.
Thanks to its highly resistant materials, the premium cap is optimally suited to utilize in processes which are both chemically and thermally demanding. Due to its high temperature resistance, it is ideal for demanding processes, such as hot-air / dry-heat sterilisation and depyrogenation at 200°C. The materials used meet the requirements of the FDA/USP/EP and JP. Consequently the premium cap is particularly suitable for use in the pharmaceutical industry. Due to the PTFE-coated silicone cap liner, the premium cap provides a reliable and tight seal. Therefore it is ideal for the long term storage and transport of sensitive products. The matching PFA pouring ring permits drip-free pouring is available separately. Replacement PTFE faced silicone cap liners are available (platinum catalysed silicone)
  • High purity polymer without any colourants
  • USP Class VI Pharmaceutical grade materials
  • Fluoropolymer cap offers high temperature resistance
  • Fluoropolymer cap offers excellent chemical resistance
  • High tightness due to PTFE-coated silicone cap liner seal
  • Fully compatible with GL 25 or 45 threads and therefore highly versatile
  • The matching PFA pouring ring permits drip-free pouring and replacement PTFE faced silicone cap liners (platinum-cured silicone) are available separately
TRUSTED_SUSTAINABILITY
Specifications
Description Screw cap
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Reservoir cap for supplying laboratory instruments; Port assemblies for spinner flasks, and cell culture vessels; Single port vent for autoclaving media in large bottles; Transport and shipping
Material PFA, PTFE, Silicone
Quantity 5/Pk.
Thread Size (Metric) GL 25
Diameter (Metric) 32 mm
Autoclavable Autoclavable
Product Content Correction
Product Title
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.