Learn More
DWK Life Sciences DURAN™ Liebig Condenser, (West Condenser), with 2 standard ground joints, and 2 screw-on plastic hose connections
Product Code. 10756921 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No:
NS 14/23
NS 24/29
NS 29/32
Length (Metric):
160 mm
250 mm
400 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Unit Size
10756921 NS 14/23 160 mm Each
10325441 NS 29/32 250 mm Each
10243171 NS 14/23 250 mm Each
10365391 NS 24/29 250 mm Each
10480493 NS 24/29 400 mm Each
10080051 NS 29/32 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10756921 Supplier DWK Life Sciences Supplier No. 242516107
  • ONLINE EXCLUSIVE PRICE
    57.55€
    50.32€ / Pack of 1
    Save 7.23€
    12% Off
Estimated Shipment: 22-12-2026
The Liebig condenser is suitable for condensing vapours from reaction mixtures (distillation).
The Liebig condenser is one of the best-known types of condenser and is used for distilling reaction mixtures. The condenser is a tube with standard ground ends and a glass jacket. The cooling water is fed through the jacket in counter flow principle to increase the cooling capacity.
  • Relatively small heat exchange surface and thus relatively low cooling capacity
  • Two removable plastic olives for inlet and outlet of the cooling medium
  • Tight closure due to standard ground joint
Specifications
For Use With (Application) Product condenser for distillate separations; Distillation; Countercurrent cooling; Condestandard ground jointers of length 250 mm (with standard ground joint 14/23, standard ground joint 24/29 and standard ground joint 29/32) and 400 mm (with standard ground joint 24/29) are special sizes and not according to DIN
Length (Metric) 160 mm
Quantity 1 Ea.
Standard Taper Joint No NS 14/23
Material Borosilicate Glass (3.3), PBT, Polypropylene, Silicone