Få mere at vide
Få mere at vide
DWK Life Sciences DURAN™ High Temperature Screw Cap, PTFE-Coated, Peroxide cured silicone, Red PBT
Shop alle DWK Life Sciences produkter
Klik for at se tilgængelige muligheder
Thread Size (Metric):
GL 14
GL 18
GL 25
GL 32
GL 45
Pakningsstørrelse:
Pack of 10
Beskrivelse
The red polybutylene terephthalate (PBT) GL cap and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. Screw cap seal has a PTFE faced cap liner for efficient sealing. ETFE pouring ring ensures drip free pouring.
- Excellent chemical resistance due to PTFE coating
- Cap liner forms a tight seal with bottle Peroxide catalysed silicone
- Peroxide catalysed silicone
- Suitable for PBT screw caps and tamper-evident screw caps
- Fully compliant with current international food contact regulations
Tekniske data
Tekniske data
| Color | Red |
| Description | Screw cap |
| For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Suitable for dry heat sterilization; Pressure applications; Storage of corrosive reagents; Chemical and thermal reactions; Long term / archival storage of samples, media and reagents; Sampling; Transport and shipping |
| Material | PBT, ETFE, Fiberglass |
| Quantity | 10/Pk. |
| Screw Material | Polybutylene Terephthalate (PBT) with glass fiber |
| Thread Size (Metric) | GL 14 |
| Liner Material | PTFE-coated Silicone |
| Closure Liner | PTFE/Silicone |
| Closure Material | PBT with Glass Fiber |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion