Learn More
DWK Life Sciences DURAN™ Hexagonal Stopper, pointed, diamond-ground
Shop All DWK Life Sciences Products
Click to view available options
Size:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Unit Size:
Each
Pack of 10
Pack of 100
Description
DURAN™ hexagonal hollow plugs are used for sealing laboratory equipment such as volumetric flasks, mixing cylinders, ground joint flasks, etc. DURAN™ hexagonal hollow plugs are available in 2 standard versions - pointed - flat - Sizes analogous to DURAN™ conical ground joints. Cut quality: DG = Diamond Ground
- Pointed
- Diamond-ground
- With a dripping syringe, for example, a condensate can be collected and the reaction medium recirculated.
- hollow glass stopper
Specifications
Specifications
| For Use With (Application) | Closure of vessels with standard ground joints |
| Quantity | 100/Pk. |
| Size | NS 14/23 |
| Autoclavable | Autoclavable |
| Material | Borosilicate Glass |