Learn More
DWK Life Sciences DURAN™ Glass Stopper, Amber, Standard Taper Ground Joint, Octagonal flat head
Amber DURAN™ glass ground stopper.
Brand: DWK Life Sciences 216271203
24.13 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.15300kg
Description
Semi-hollow or solid amber DURAN™ glass stoppers can be used to close glassware items such as flasks, separatory funnels or reagent bottles with ground joint necks. The Standard Taper ground glass joint forms a tight, leak-proof seal. Use grease for heating or vacuum applications. With octagonal shaped head. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!
- USP/EP UV light protection due to external amber colouration
- Very durable and chemically resistant amber colour
- All USP Type 1 glass components and therefore suitable for storage of pharmaceutical API materials
- All USP glass components and therefore suitable for storage of corrosive and aggressive materials
- Tight closure due to standard taper ground joint
- Octagonal shaped head
Specifications
Semi-hollow | |
1 Ea. | |
ST 45/40 |
Storage of light sensitive reagents, pharmaceuticals, and chemicals; Storage of air sensitive reagents, and chemicals; Storage of hygroscopic solids, granules and powders; Storage of pharmaceutical APIs; Sealing laboratory glassware such as flasks, reagent bottles, and mixing cylinders | |
Borosilicate Glass | |
No |