Learn More
Lenz Laborglasinstrumente™ Double Walled Coil Condensers with GL Threads
Click to view available options
Standard Taper Joint No:
14/23
29/32
Length (Metric) Jacket:
160 mm
400 mm
Unit Size:
Each
Description
- Acc. to DIN 12593
- With NS socket and cone
- Corrosion-free and resistant to thermal shock
- Water connection: GL 14 threads (with two threaded plastic olives)
Specifications
Specifications
| Certifications/Compliance | DIN 12593 and in addition to DIN |
| For Use With (Application) | For condensing vapors |
| Length (Metric) Jacket | 400 mm |
| Quantity | 1 Each |
| Standard Taper Joint No | 29/32 |
| Material | Duran Glass |
| Type | Coil Condenser |