Learn More
Lenz Laborglasinstrumente™ Duran Glass Columns acc. to Hempel
39.70€ - 263.94€
Product Code | Brand | Length (Metric) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Includes | Price | Quantity & Availability | ||||
10722342
|
Lenz Laborglasinstrumente™
05115201 |
200mm | NS socket and NS cone |
39.70€
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
10539751
|
Lenz Laborglasinstrumente™
05119304 |
300mm | Fixed vacuum jacket |
263.94€
Each |
Estimated Shipment: 28-05-2024
Log in to see stock. |
||||
10050511
|
Lenz Laborglasinstrumente™
05117304 |
300mm | NS socket and NS cone and Removable Glass jacket |
122.96€
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
10057570
|
Lenz Laborglasinstrumente™
05115504 |
500mm | NS socket and NS cone |
71.60€
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
Specifications
Columns acc. to Hempel | |
Duran Glass | |
1Each |