Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Coil Condenser
Product Code. 10695031
Change view
Click to view available options
Standard Taper Joint No:
NS 14/23
NS 24/29
NS 29/32
Length (Metric) Jacket:
160 mm
250 mm
400 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Jacket Unit Size
10695031 NS 29/32 250 mm Each
10395781 NS 14/23 160 mm Each
10204391 NS 24/29 250 mm Each
10003922 NS 29/32 160 mm Each
10732342 NS 29/32 400 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10695031 Supplier Lenz Laborglasinstrumente™ Supplier No. 05250304
  • ONLINE EXCLUSIVE PRICE
    175.00€
    110.49€ / Pack of 1
    Save 64.51€
    36% Off
Estimated Shipment: 11-12-2026
Duran™ Glass Coil Condensers made of DURAN™ tubing.
  • Duran™ Glass
Specifications
Certifications/Compliance DIN 12593 and in addition to DIN
Description Cooling Surface: 400 cm2
For Use With (Application) For condensing vapors
Length (Metric) Jacket 250 mm
Quantity 1 Ea.
Standard Taper Joint No NS 29/32
Material Duran Glass
Type Coil Condenser