Learn More
DWK Life Sciences DURAN™ Gas Distribution Tube
Product Code. 15242059 Shop All DWK Life Sciences Products
Change view
Click to view available options
Porosity:
0
1
2
Diameter (Metric) Outside:
9 mm
11 mm
Unit Size:
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Porosity Diameter (Metric) Outside Unit Size
15242059 1 11 mm Pack of 5
15212059 0 9 mm Pack of 5
15222059 1 9 mm Pack of 5
15232059 2 9 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15242059 Supplier DWK Life Sciences Supplier No. 258562106
  • ONLINE EXCLUSIVE PRICE
    331.00€
    288.43€ / Pack of 5
    Save 42.57€
    12% Off
Estimated Shipment: 04-03-2027
Gas is introduced into a liquid via the gas distribution tube for cleaning. The molten glass filter plate produces as small gas bubbles as possible and thus a large contact surface with the liquid.
For gas filtration and cleaning, gas washing bottles are required which consist of a gas distribution tube, a suitable bottle and a gas outlet opening. In this test setup, the gas is forced to flow through a liquid before it is released. The gas distribution pipe is immersed in the liquid for this purpose, the gas interferes through it and impurities of the gas are bound in the liquid. The gas is cleaned or washed so to speak. A filter disk in the gas distribution tube creates air bubbles that are as small as possible and thus form as large a contact area as possible.
  • The introduced gas flow is divided into small bubbles by means of a built-in glass distribution tube
  • Larger contact surface due to small bubbles of the gas flow
  • Fired-on, highly durable white print
Specifications
Material Borosilicate Glass (3.3)
Type Center-Mounted Filter Bowl
Porosity 1
Diameter (Metric) Outside 11 mm
For Use With (Application) Reactions between gases and liquids; Purification of gas mixtures; Filtering; Version with side or centrally mounted filter cup available
Quantity 5/Pk.