Learn More
DWK Life Sciences DURAN™ Flask, Kjeldahl
Product Code. 11737981 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
750 mL
1000 mL
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11737981 750 mL Pack of 10
11717971 100 mL Pack of 10
11727971 250 mL Pack of 10
15221669 500 mL Pack of 10
15231669 1000 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11737981 Supplier DWK Life Sciences Supplier No. 212315109
  • ONLINE EXCLUSIVE PRICE
    345.45€
    337.05€ / Pack of 10
    Save 8.40€
    2% Off
Estimated Shipment: 12-10-2026
Add to basket
Add to basket
For nitrogen determination.
DURAN™ Kjeldahl flasks are subjected to extreme chemical and physical stresses. Therefore, the wall thickness should not be too thick to avoid overheating, which can lead to breakage due to remaining (permanent) stresses, on the one hand, and not too thin due to the noticeable glass removal in the alkaline medium, on the other hand. In order to offer the user the safest possible compromise while taking the technological conditions into account, the wall thickness of the DURAN™ Kjeldahl flasks in the body are 1-2 mm.
  • Wall thickness adapted to the application (glass removal by alkaline medium taken into account)
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Base 115 mm
Capacity (Metric) 750 mL
Material Borosilicate Glass (3.3)
For Use With (Application) For nitrogen determination
No. per Pack 10/Pk.