Learn More
DWK Life Sciences DURAN™ Dropping Funnel, cylindrical, with scale
Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Description
The dosed addition of solutions, reaction mixtures and/or chemicals is ideally carried out using a dropping funnel.
- With standard ground stopcock and retaining device
- The rate of supply can be adjusted
- Cylindrical with scale
- Tight closure due to standard ground joint
- Fired-on, highly durable white print
- Easy-to-read scale with graduation marks to estimate the content
Specifications
Specifications
| Diameter (Metric)Top | 13 mm |
| Capacity (Metric) | 1000 mL |
| Material | Boro 3.3, Polypropylene |
| Autoclavable | Autoclavable |
| For Use With (Application) | Uniform and metered liquid supply to a reaction mixture; Measuring liquid amounts |
| Graduations | 20 mL |
| Quantity | 1 Ea. |