Learn More
Hirschmann™ DURAN™ Dafert Pattern Burettes with Blue Graduations
Product Code. 13198009
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
13198009 25 mL Pack of 2
13108019 50 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13198009 Supplier Hirschmann™ Supplier No. 3330170
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Excellent for dispensing measured amounts of chemical solutions in the laboratory.
  • Schellbach
  • Short line
  • With blue graduation
  • Three-way stopcock with glass key
  • Straight form
Specifications
Capacity (Metric) 25 mL
Material Glass
Graduations 0.05 mL
Includes Schellbach Stripe, Filling Funnel
Scale Blue
Stopcock Straight
Stopcock Material Glass
Tolerance ±0.045 mL
Type Dafert Pattern Buret
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Sekretesspolicy.