Learn More
DWK Life Sciences DURAN™ Cylinder, with knobbed lid, polished rim
Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
1000 mL
12,000 mL
2000 mL
250 mL
500 mL
6000 mL
Unit Size:
Each
Pack of 10
Description
Ideally, containers that can be closed are used to store substances and materials. In this way, substances and materials can be protected from unwanted effects. These containers are available in various shapes such as cans, cylinders or jars. The choice of the vessel depends on the application and/or nature of the material.
- The lid can be opened easily while using the button
- Fired-on, highly durable white ceramic
Specifications
Specifications
| Material | Borosilicate Glass (3.3) |
| Diameter (Metric) Outside | 120 mm |
| For Use With (Application) | Storage and transport |
| Capacity (Metric) | 1000 mL |
| Autoclavable | Autoclavable |
| Quantity | 1 Ea. |