Learn More
DWK Life Sciences DURAN™ Crystallizing Dish, without spout
Product Code. 11766582 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
20 mL
40 mL
60 mL
100 mL
150 mL
300 mL
500 mL
900 mL
2000 mL
3500 mL
Unit Size:
Pack of 10
Product Code. 11766582
Brand: DWK Life Sciences 213133408
Only null left
Add to basket
Add to basket
The flat shape as well as the flat bottom of the crystallizing dish allow an even evaporation of solvents and thus the crystallization of solids from solutions and suspensions.
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work.
  • Flat and wide shape allow solvents to evaporate quickly and evenly
  • Large ground base
  • Spout for clean pouring
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Capacity (Metric) 60 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Crystallization; Preparation of concentrated solutions; Available with spout DIN 12 338 or without spout DIN 12 337
No. per Pack 10/Pk.
Diameter (Metric) 60 mm