Learn More
DWK Life Sciences DURAN™ Coil Distillate Condenser, with 2 standard ground joints, and 2 screw-on plastic hose connections
Product Code. 10528821 Shop All DWK Life Sciences Products
Change view
Click to view available options
Standard Taper Joint No:
NS 24/29
NS 29/32
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Unit Size
10528821 NS 24/29 Each
10080011 NS 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10528821 Supplier DWK Life Sciences Supplier No. 242537105
  • ONLINE EXCLUSIVE PRICE
    122.85€
    106.65€ / Pack of 1
    Save 16.20€
    13% Off
Estimated Shipment: 23-10-2026
The coil condenser serves to distill a reaction mixture or to cool a condensate.
Typical for the coil condenser is its spiral riser tube, which is located in a jacket and thus washed around by the cooling medium (contrary to the Dimroth cooler). Due to this condensate flow, the condensation performance is very low although the cooling section is very long. This type of condenser is therefore usually used as a downstream additional condenser and not as an independent distillation condenser. Due to the better drainage of condensate in the convoluted riser tube, the condenser must be installed in a vertical position.
  • Usually used as a downstream additional cooler due to long cooling section
  • Two removable plastic olives for inlet and outlet of the cooling medium
  • Tight closure due to standard ground joint
Specifications
For Use With (Application) Condensate cooling; Product condenser for distillate separations; Distillation
Quantity 1 Ea.
Standard Taper Joint No NS 24/29
Material Borosilicate Glass (3.3), PBT, Polypropylene, Silicone