Learn More
Hirschmann™ DURAN™ Burette Stopcock Lateral
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
Burette 2 to 10 mL
Burette 25 mL
Burette 50 mL
missing translation for 'unitSize'
Each
Description
- With glass key
Specifications
Specifications
| Capacity (Metric) | 25 mL |
| For Use With (Equipment) | Burette 25 mL |
| Material | Glass |
| Includes | Glass Key |
| Stopcock | Lateral |
| Type | Buret |