Learn More
Hirschmann™ DURAN™ Burette Stopcock Lateral
Click to view available options
For Use With (Equipment):
Burette 2 to 10 mL
Burette 25 mL
Burette 50 mL
Unit Size:
Each
Description
- With glass key
Specifications
Specifications
| Capacity (Metric) | 10 mL |
| For Use With (Equipment) | Burette 2 to 10 mL |
| Material | Glass |
| Includes | Glass Key |
| Stopcock | Lateral |
| Type | Buret |