Learn More
DWK Life Sciences DURAN™ Burette Stopcock Key, side entry, drilled and ground, with three part retaining device
Product Code. 15824518 Shop All DWK Life Sciences Products
Change view
Click to view available options
No. per Pack:
50/Pk.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Pack Unit Size
15824518 50/Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15824518 Supplier DWK Life Sciences Supplier No. 286035504
  • ONLINE EXCLUSIVE PRICE
    730.00€
    634.61€ / Pack of 50
    Save 95.39€
    13% Off
Estimated Shipment: 29-01-2027
For use in burettes.
Lateral DURAN™ burette tap Plug with 3-piece tap locking have a calibrated tip (Øi 0.80-0.85 mm).
  • For the lateral burette outlet e.g. for automatic burettes
  • Burette stopcock key with calibrated tip (Øi 0,80-0,85 mm) for accurate working
  • Ground tap plug, with 3-part tap safety catch
Specifications
Plug Bore 2.5 mm
Material Borosilicate Glass (3.3), Polypropylene
Autoclavable Autoclavable
Closure Size 12.5 NS
No. per Pack 50/Pk.
For Use With (Application) Installation in glass apparatus and devices