Learn More
DWK Life Sciences DURAN™ Burette Stopcock, angled, complete with glass key
Shop All DWK Life Sciences Products
Click to view available options
Plug Bore:
2.5 mm
Unit Size:
Pack of 50
Description
Lateral DURAN™ burette taps complete with threaded tap plugs for attaching to burettes with calibrated tip (Øi 0.80-0.85 mm).
- Side burette tap case with glass plug
- Tip not calibrated
- Glass articles for further processing
Specifications
Specifications
| Plug Bore | 2.5 mm |
| Diameter (Metric) | 8 mm |
| Material | Borosilicate Glass (3.3), Polypropylene |
| Autoclavable | Autoclavable |
| Closure Size | 12.5 NS |
| Width (Metric) | 1.5 mm |
| No. per Pack | 50/Pk. |
| For Use With (Application) | For attaching to burettes |