Learn More
DWK Life Sciences DURAN™ Blank for reaction vessel
Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
10,000 mL
1000 mL
2000 mL
4000 mL
6000 mL
missing translation for 'unitSize'
Each
Description
DURAN™ blanks for reaction vessels are convincing due to their good wall thickness distribution and enable the cost-effective production of reaction vessels with reproducible geometry. For pressure resistance, please refer to our notes in the technical appendix of the laboratory glassware catalogue. For glass articles which deviate from the original form due to further processing, the permissible operating data must be determined in accordance with the technical rules and regulations.
- Component for further processing into reaction vessels
- Due to the geometry and the massive wall thickness, the blanks are suitable for devices with pressure and vacuum applications.
- Glass articles for further processing
Spécification
Spécification
| Diameter (Metric) Base | 200 mm |
| Capacity (Metric) | 4000 mL |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | For processing as a reaction vessel |
| No. per Pack | 1/Pk. |
| Autoclavable | Autoclavable |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu