Learn More
H-B Instrument™ DURAC™ Digital Stopwatches
Product Code. 17673175
Change view
Click to view available options
Description:
With Clock and Calendar
With Clock, Alarm and Calendar
Time Range:
10 hr.
Chronograph: 1/100 sec
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Time Range Unit Size
17673175 With Clock, Alarm and Calendar Chronograph: 1/100 sec Each
17663175 With Clock and Calendar 10 hr. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17673175 Supplier H-B Instrument™ Supplier No. B617005300
  • ONLINE EXCLUSIVE PRICE
    53.80€
    22.60€ / Pack of 1
    Save 31.20€
    57% Off
Estimated Shipment: 03-02-2027
Supplied with multi-language (English, French, German, Italian, Portuguese and Spanish) instructions, these electronic stopwatches include a calendar and a clock.
  • Features Certificate of Calibration and individual serial number
  • Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
  • B61700-5200 stores 30 lap times and calculates lap time average, allows for 99 split lap times, and has a 10/320SEC pacer
  • B61700-5300 will store 1 lap time and includes an alarm
  • Requires one CR2032 battery (included)
Specifications
Product Type Stopwatch
Time Range Chronograph: 1/100 sec
Accuracy 0.01 sec
Resolution 0.001
Description With Clock, Alarm and Calendar
Width (English) 2.5 in.
Width (Metric) 63 mm
Height (English) 3.1 in.
Height (Metric) 80 mm