Learn More
Duloxetine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Duloxetine hydrochloride |
| CAS | 136434-34-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H19 N O S . Cl H |
| Quantity | 100 mg |
| Synonym | 2-Thiophenepropanamine, N-methyl-γ-(1-naphthalenyloxy)-, hydrochloride (1:1), (γS)-, 2-Thiophenepropanamine, N-methyl-γ-(1-naphthalenyloxy)-, hydrochloride, (S)-, 2-Thiophenepropanamine, N-methyl-γ-(1-naphthalenyloxy)-, hydrochloride, (γS)- (9CI), (S)-(+)-Duloxetine hydrochloride, (S)-(+)-N-Methyl-3-(1-naphthalenyloxy)-3-(2-thienyl)propanamine hydrochloride, (S)-Duloxetine hydrochloride, (S)-N-Methyl-3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propan-1-amine hydrochloride, (S)-γ-(1-Naphthalenoxy)-N-methylthiophene-2-propanamine monohydrochloride, Cymbalta, Dulane 20, Duloxetine hydrochloride, LY 248686 HCl, (γS)-N-Methyl-γ-(1-naphthalenyloxy)-2-thiophenepropanamine hydrochloride |
| InChI Formula | InChI=1S/C18H19NOS.ClH/c1-19-12-11-17(18-10-5-13-21-18)20-16-9-4-7-14-6-2-3-8-15(14)16;/h2-10,13,17,19H,11-12H2,1H3;1H/t17-;/m0./s1 |
| SMILES | Cl.CNCC[C@H](Oc1cccc2ccccc12)c3cccs3 |
| IUPAC Name | (3S)-N-methyl-3-naphthalen-1-yloxy-3-thiophen-2-ylpropan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 333.88 |
| Show More |