missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Dulbecco's Modified Eagle Medium Low Glucose

Product Code. 31908445 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31908445 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908445 Supplier Sartorius Supplier No. 010501A
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DMEM is a widely used basal medium for supporting the growth of many different mammalian cells.

  • Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells.
  • A variety of DMEM modifications are available for a range of cell culture applications.
  • Modified as follows:
    • With Glucose
    • With Sodium Pyruvate
    • Without L-Glutamine

Specifications

Classification Serum-free
Form Liquid
Product Type DMEM Cell Culture Media
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Glucose, Sodium Pyruvate
Without Additives No Glutamine
Quantity 500 mL
Culture Type Mammalian Cell Culture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.