Learn More
3M™ Dual Lock™ Reclosable Fastener Twin Pack SJ3540
Product Code. 17217800 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
2.5 m x 25 mm
Unit Size:
4 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17217800 2.5 m x 25 mm 4 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17217800 Supplier 3M™ Supplier No. 7100123339
  • ONLINE EXCLUSIVE PRICE
    267.00€
    148.74€ / 4 rolls
    Save 118.26€
    44% Off
Estimated Shipment: 16-12-2026
Reclosable Fastener Twin Pack
  • Black fastener with mushroom shaped stems, backed with a rubber based pressure sensitive adhesive
  • Type 250 mates to Type 170, 250 or 400. Closes with an audible snap
  • Designed to bond to many plastics, powder coated paints and other low surface energy materials
  • Opens and closes 1000 times before losing 50% of its original tensile strength
  • High initial tack
  • Suitable for indoor applications
TRUSTED_SUSTAINABILITY
Specifications
Format Roll
Color Black
Dimensions (L x W) 2.5 m x 25 mm