Learn More
Trajan Scientific and Medical™ Dual Gauge Spare Needles
Click to view available options
Needle Gauge:
23 and 26 G
missing translation for 'unitSize'
Pack of 2
Description
- Dual Gauge: 23, 26
- Gas Tight
- 0.63/0.47mm OD
Specifications
Specifications
| Needle Gauge | 23 and 26 G |
| Diameter (Metric) Outer | 0.47 to 0.63 mm |
| Diameter (Metric) Outer Needle | 0.47 to 0.63 mm |
| Product Type | Replacement Needle |
| Volume (Metric) | 10 μL |
| Needle Point Style | Cone |
| For Use With (Equipment) | Syringes (10μL) |