Learn More
Tocris Bioscience™ DSR 6434
Potent TLR7 agonist
Brand: Tocris Bioscience™ 4809/10
This item is not returnable.
View return policy
Description
DSR 6434 is a potent TLR7 agonist (EC50 = 7.2 nM). Activates several immune effector cells in vitro. Suppresses metastasis in vivo and enhances efficacy of radiation therapy in a model of colorectal carcinoma.Specifications
| DSR 6434 | |
| 6-Amino-2-(butylamino)-9-[[6-[2-(dimethylamino)ethoxy]-3-pyridinyl]methyl]-7,9-dihydro-8H-purin-8-one | |
| Soluble to 50mM in 1eq. HCl and to 100mM in DMSO | |
| Bioactive Small Molecule |
| 1059070-10-8 | |
| 10 mg | |
| Solid |
Product Title
Spot an opportunity for improvement?
Research Use Only. Not For Human or Veterinary Use