missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DSCR1 (Human) Recombinant Protein
Human DSCR1 full-length ORF ( AAH02864, 1 a.a. - 197 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001827-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MEEVDLRDLPSATIACHLDPRVFVDGLCRAKFESLFRTYDKDITFQYFKSFKRVRINFSNPFSAADARLQLHKTEFLGKEMKLYFAQTLHIGSSHLAPPNPDKQFLISPPASPPVGWKQVEDATPVINYDLLYAISKLGPGEKYELHAATDTTPSVVVHVCESDQEKEEEEEMERMRRPKPKIIQTRRPEYTQIHLS
Specifications
AAH02864 | |
regulator of calcineurin 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RCAN1 | |
GST |
1827 | |
Wheat germ expression system | |
10 μg | |
ADAPT78, CSP1, DSC1, DSCR1, MCIP1, RCN1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |