missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DS2
Positive allosteric modulator of δ-containing GABAA receptors
246.79€ - 988.25€
Specifications
Purity | >97% |
---|---|
Product Type | DS2 |
Molecular Weight (g/mol) | 353.83 |
Inhibitors | DS2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18005602
|
Tocris Bioscience™
3679/10 |
10 mg |
246.79€
10mg |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
18035312
|
Tocris Bioscience™
3679/50 |
50 mg |
988.25€
50mg |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
Description
Positive allosteric modulator of δ-subunit containing GABAA receptors (EC50 = 142 nM for α4β3δ receptors). Displays no effects on GABA responses mediated by α4β3γ2 and α1β3γ2 receptors.Specifications
>97% | |
DS2 | |
353.83 | |
DS2 |
For Research Use Only